Order Entry
Belgium
ContactUsLinkComponent
 
OMNIMMP® RED Fluorogenic Substrate
Catalog # ENZOBMLP2770100
Supplier:  Enzo Life Sciences
CAS Number:  
OMNIMMP® RED Fluorogenic Substrate
Catalog # ENZOBMLP2770100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Storage conditions:
    −20 °C
  • Protein/peptide name:
    RED fluorogenic substrate
  • Purity:
    >90% (HPLC)
  • Solubility:
    Soluble in DMSO (50 mM)
  • Molecular weight:
    1910.7
  • Sequence:
    TQ3-GABA-Pro-Cha-Abu-Smc-His-Ala-Dab(6'-TAMRA)-Ala-Lys-NH₂
  • Formulation:
    Lyophilised

Specifications

About this item

Highly-quenched, ultra-bright fluorogenic substrate for most MMPs. 6'-TAMRA fluorescence is thoroughly quenched by the TQ3 group until cleavage by MMPs separates the two moieties. The OMNIMMP® RED substrate offers key advantages over other MMP substrates.