PAP-AMC, Tyrosinase substrate ≥98% (by TLC)
Catalog # ENZOBMLEI4050100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:0,1 mg
- Seq:1286
Specifications
Specification Test Results
- Purity:≥98% (TLC)
Specification Test Results
About this item
:
:310.3 g/mol
Melting Pt:238…242 °C
Storage Temperature:Freezer
Formula:C₁₇H₁₄N₂O₄