Order Entry
Belgium
ContactUsLinkComponent
4-Diphenylacetoxy-N-methylpiperidine methiodide ≥98% (by HPLC)
4-Diphenylacetoxy-N-methylpiperidine methiodide ≥98% (by HPLC)
  ENZOBMLC1040050
 :  
undefined
4-Diphenylacetoxy-N-methylpiperidine methiodide ≥98% (by HPLC)
  ENZOBMLC1040050
 :  

 

  • Pk:
    50 mg
  • Seq:
    2370

 

 

  • Purity:
    ≥98% (HPLC)

 

 

 :
 :451.3 g/mol
 :Ambient
 :1952-15-4
 :C₂₁H₂₆INO₂
 :MFCD00078564