Order Entry
Belgium
ContactUsLinkComponent
 
DNA (Deoxyribonucleic acid) (from calf thymus), purified
Catalog # ENZOALX840040C010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
DNA (Deoxyribonucleic acid) (from calf thymus), purified
Catalog # ENZOALX840040C010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Seq:
    1106

Specifications

About this item

 :
Storage Temperature:Refrigerator
CAS Number:9007-49-2
 :73049-39-5
MDL Number:MFCD00130922