Order Entry
Belgium
ContactUsLinkComponent
Lucigenin ≥97% (by HPLC)
Lucigenin ≥97% (by HPLC)
Catalog # ENZOALX620061M050
Supplier:  Enzo Life Sciences
CAS Number:  
Lucigenin ≥97% (by HPLC)
Catalog # ENZOALX620061M050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :510.51 g/mol
CAS Number:2315-97-1
 :22103-92-0
Formula:C₂₈H₂₂N₄O₆
MDL Number:MFCD00011925