DL-α-Tocopherol (Vitamin E)
Catalog # ENZOALX460018G001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:430.71 g/mol
EINECS:233-466-0
CAS Number:10191-41-0
:1406-66-2
Formula:C₂₉H₅₀O₂
MDL Number:MFCD00072051