Order Entry
Belgium
ContactUsLinkComponent
Calcium pantothenate ≥98% (by HPLC)
Calcium pantothenate ≥98% (by HPLC)
Catalog # ENZOALX460003G010
Supplier:  Enzo Life Sciences
CAS Number:  
Calcium pantothenate ≥98% (by HPLC)
Catalog # ENZOALX460003G010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%
  • Identity:
    Determined by IR
  • Appearance:
    White to off-white powder

Specification Test Results

About this item

 :
 :476.54 g/mol
Merck Index:14,07015
Storage Temperature:Ambient
EINECS:205-278-9
CAS Number:137-08-6
Formula:C₁₈H₃₂CaN₂O₁₀
MDL Number:MFCD00002766