Order Entry
Belgium
ContactUsLinkComponent
Sapintoxin D (high purity) ≥98% (by HPLC)
Toxic
Irritant
Health
Sapintoxin D (high purity) ≥98% (by HPLC)
Catalog # ENZOALX445047M005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Toxic
Irritant
Health
Sapintoxin D (high purity) ≥98% (by HPLC)
Catalog # ENZOALX445047M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%

Specification Test Results

About this item

 :
 :539.6 g/mol
Storage Temperature:Freezer
UN:3462
ADR:6.1,I
 :GHS06
 :GHS07
 :GHS08
CAS Number:80998-07-8
Formula:C₃₀H₃₇NO₈