Demecolcine ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:477-30-5
:
Formula:C₂₁H₂₅NO₅
MDL Number:MFCD00075459
:371.433 g/mol
Frequently Bought Together
Cellulose chromatography papers, Whatman™
Supplier: Whatman products (Cytiva)
Tube 1,5ml with white screw cap non-skirted sterile 1 * 2.500 items
Supplier: Greiner Bio-One
Cryoboxes B51/B52/B63/B64/B74/B94-series
Supplier: GLW
Bioreactors, Wheaton® CELLine™
Supplier: DWK Life Sciences
BASE PLATE - TOPAS REPLACEMENT PART 1 * 1 Pce
Supplier: MARKET SOURCE PART PROCESS
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
VWR® Safe-T-Flex, Caps for Test Tubes
Supplier: VWR Collection
Detectable scoops, SteriPlast®
Supplier: BURKLE
Eppendorf twin.tec® PCR Plates 96-well, 150 µl
Supplier: EPPENDORF
96-Well microplates, non treated, BRANDplates®
Supplier: Brand
Sampling spoon, long handle, disposable Bio
Supplier: BURKLE
Serological pipettes, Sterilin®
Supplier: STERILIN
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES