Order Entry
Belgium
ContactUsLinkComponent
 
(-)-p-Bromotetramisole oxalate ≥98% (by HPLC)
Catalog # ENZOALX430025M005
Supplier:  Enzo Life Sciences
CAS Number:  
(-)-p-Bromotetramisole oxalate ≥98% (by HPLC)
Catalog # ENZOALX430025M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :373.22 g/mol
Storage Temperature:Refrigerator
CAS Number:62284-79-1
Formula:C₁₁H₁₁BrN₂S·C₂H₂O₄
MDL Number:MFCD00070406