Order Entry
Belgium
ContactUsLinkComponent
 
 
 
AM 281 ≥98% (by HPLC)
  ENZOALX270240M005
 :  
 
 
AM 281 ≥98% (by HPLC)
  ENZOALX270240M005
 :  

 

  • Pk:
    5 mg
  • Seq:
    2210

 

 

  • Purity:
    ≥98% (HPLC)
  • Identity:
    Determined by ¹H-NMR

 

 

 :
 :557.2 g/mol
 :Refrigerator
 :2811
 :6.1,III
 :GHS06
 :GHS07
 :202463-68-1
 :C₂₁H₁₉Cl₂IN₄O₂