Order Entry
Belgium
ContactUsLinkComponent
2-tert-Butylhydroquinone
2-tert-Butylhydroquinone
Catalog # ENZOALX270061G010
Supplier:  Enzo Life Sciences
CAS Number:  
2-tert-Butylhydroquinone
Catalog # ENZOALX270061G010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :166.22 g/mol
Boiling Pt:273 °C (1013 hPa)
Melting Pt:128 °C
Density:1,05 g/cm³ (20 °C)
Flash Pt:171 °C
Storage Temperature:Ambient
EINECS:217-752-2
UN:3077
ADR:9,III
CAS Number:1948-33-0
Formula:C₁₀H₁₄O₂
MDL Number:MFCD00002344