EQ1 NHS ester amine-reactive quencher dye
Catalog # ENZOENZCHM1650001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- Excitation maximum:535 nm
- Emission maximum:none
- Extinction Coefficient:39,200 M-1cm-1
- Correction Factor (260 nm):0.34
- Correction Factor (280 nm):0.27
- Purity:≥80% (HPLC)
- Appearance:Dark red solid
Specification Test Results
About this item
:
:535.5 g/mol