Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyanine 3-NHS ester chloride
Cyanine 3-NHS ester chloride, CELLESTIAL® fluorescent dye
Cyanine 3-NHS ester chloride, CELLESTIAL® fluorescent dye
ENZOENZ42541
:
ENZO LIFE SCIENCES
:
Cyanine 3-NHS ester chloride, CELLESTIAL® fluorescent dye
ENZOENZ42541
:
ENZO LIFE SCIENCES
:
Pk:
50 Nmo
Seq:
0381
:
:
590.15 g/mol
:
GHS07
:
GHS08
:
Refrigerator
:
146368-16-3
:
C₃₄H₄₀ClN₃O₄