Order Entry
Belgium
ContactUsLinkComponent
Prostaglandin A1 ≥98% (by HPLC)
Prostaglandin A1 ≥98% (by HPLC)
Catalog # ENZOBMLPG0010010
Supplier:  Enzo Life Sciences
CAS Number:  
Prostaglandin A1 ≥98% (by HPLC)
Catalog # ENZOBMLPG0010010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :336.5 g/mol
Storage Temperature:Freezer
CAS Number:14152-28-4
Formula:C₂₀H₃₂O₄