Hydroxy-2-naphthalenylmethylphosphonic acid ≥98% (by TLC)
:132541-52-7
:
:C₁₁H₁₁O₄P
:238.2 g/mol
:≥250 °C
:Freezer
Frequently Bought Together
Microplates, 96- and 384-well, Nunc™
: Thermo Fisher Scientific
VWR®, Sampling Scoops
: VWR Collection
Centrifuge tubes, SuperClear®
: LABCON
VWR®, Stoppers, PE
: VWR Collection
VWR®, Strips of PCR Tubes and Caps
: VWR Collection
Petri dishes
: Greiner Bio-One
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
: VWR Collection
Cell culture dishes, Nunc™
: Thermo Fisher Scientific
VWR® Multiwell Cell Culture Plates
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Cryobox, System 100™
: Thermo Fisher Scientific
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Transfer Pipettes, Graduated
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
: VWR Collection
Cell culture flasks, Falcon®
: Corning
Cap strips, flat top
: Thermo Fisher Scientific
DURAN® Reactor, Stirred, GLS 80 Thread
: DWK Life Sciences