Order Entry
Belgium
ContactUsLinkComponent
Zinquin ethyl ester ≥98% (by HPLC), Ultrapure fluorescent indicator, for the determination of zinc
Zinquin ethyl ester ≥98% (by HPLC), Ultrapure fluorescent indicator, for the determination of zinc
Catalog # ENZOENZ52151
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Zinquin ethyl ester ≥98% (by HPLC), Ultrapure fluorescent indicator, for the determination of zinc
Catalog # ENZOENZ52151
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :414.47 g/mol
CAS Number:181530-09-6
Formula:C₂₁H₂₂N₂O₅S