Order Entry
Belgium
ContactUsLinkComponent
H-Met-AMC ≥98% (by TLC)
H-Met-AMC ≥98% (by TLC)
Catalog # ENZOBMLP2360025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
H-Met-AMC ≥98% (by TLC)
Catalog # ENZOBMLP2360025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :420.40 g/mol
Storage Temperature:Freezer
CAS Number:94367-35-8
Formula:C₁₅H₁₈N₂O₃S·C₂HF₃O₂
MDL Number:MFCD00210065