


Pyocyanin ≥98% (by HPLC)
Catalog # ENZOENZ53001C001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 Vial
- Seq:0032
Specifications
Specification Test Results
- Purity:≥98% (HPLC)
- Formulation:Lyophilised
Specification Test Results
About this item
:
:210.2 g/mol
:GHS05
:GHS06
:GHS07
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
CAS Number:85-66-5
Formula:C₁₃H₁₀N₂O