
Phorbol-12,13-dibutyrate ≥99% (by HPLC)
Catalog # ENZOBMLPE1350005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Purity:≥99% (HPLC)
Specification Test Results
About this item
:
:504.6 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:37558-16-0
Formula:C₂₈H₄₀O₈