Order Entry
Belgium
ContactUsLinkComponent
S-Farnesyl-L-cysteine methyl ester (synthetic) ≥98% (by TLC), colourless oil
S-Farnesyl-L-cysteine methyl ester (synthetic) ≥98% (by TLC), colourless oil
Supplier:  Enzo Life Sciences
Certificates
undefined
S-Farnesyl-L-cysteine methyl ester (synthetic) ≥98% (by TLC), colourless oil
Supplier:  Enzo Life Sciences

Specification Test Results

  • Purity:
    ≥98% (TLC)
  • Source:
    Synthetic

Specification Test Results

Frequently Bought Together