Cilostamide ≥98% (by TLC)
Supplier: Enzo Life Sciences
Jump to:
About this item
PDE inhibitor
Phosphodiesterase PDE3 inhibitor (IC50=5 nM). Blocks PKB/Akt signaling pathway by inhibition of Akt-activated PDE3B. Reverses the established effects of leptin on food intake and body weight. Blocks the leptin-induced tyrosine phosphorylation of STAT3 at the hypothalamic level.
Synonyms: N-Cyclohexyl-N-methyl-4-((2-oxo-1,2-dihydroquinolin-6-yl)oxy)butanamide
- CAS Number:68550-75-4
- Formula:C₂₀H₂₆N₂O₃
- MDL Number:MFCD00673958
- Molecular Weight:342.43 g/mol
2 Options Available Below
Product Details & Documents
PDE inhibitor
Phosphodiesterase PDE3 inhibitor (IC50=5 nM). Blocks PKB/Akt signaling pathway by inhibition of Akt-activated PDE3B. Reverses the established effects of leptin on food intake and body weight. Blocks the leptin-induced tyrosine phosphorylation of STAT3 at the hypothalamic level.
Synonyms: N-Cyclohexyl-N-methyl-4-((2-oxo-1,2-dihydroquinolin-6-yl)oxy)butanamide
- CAS Number:68550-75-4
- Formula:C₂₀H₂₆N₂O₃
- MDL Number:MFCD00673958
- Molecular Weight:342.43 g/mol



