Order Entry
Belgium
ContactUsLinkComponent
Calcein AM ≥95% (by HPLC), Ultrapure
Calcein AM ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52002
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Calcein AM ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52002
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :994.86 g/mol
Storage Temperature:Freezer
CAS Number:148504-34-1
Formula:C₄₆H₄₆N₂O₂₃
MDL Number:MFCD05861516