Order Entry
Belgium
ContactUsLinkComponent
4-Phenylchalcone oxide (synthetic) ≥98% (by HPLC)
4-Phenylchalcone oxide (synthetic) ≥98% (by HPLC)
Catalog # ENZOBMLEI1080100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
4-Phenylchalcone oxide (synthetic) ≥98% (by HPLC)
Catalog # ENZOBMLEI1080100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :300.357 g/mol
Storage Temperature:Freezer
CAS Number:82389-34-2
Formula:C₂₁H₁₆O₂
MDL Number:MFCD00211155