Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Anisomycin
Anisomycin ≥97% (by TLC)
Anisomycin ≥97% (by TLC)
Anisomycin ≥97% (by TLC)
Catalog # ENZOBMLST1020050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Anisomycin ≥97% (by TLC)
Catalog # ENZOBMLST1020050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
50 mg
Seq:
2370
Specifications
About this item
Product Details
:
:
265.31 g/mol
CAS Number:
22862-76-6
Formula:
C₁₄H₁₉NO₄
MDL Number:
MFCD00077650
Product Details