Order Entry
Belgium
ContactUsLinkComponent
N-Methyl-D-aspartic acid ≥98% (by TLC)
N-Methyl-D-aspartic acid ≥98% (by TLC)
Catalog # ENZOALX105009M050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
N-Methyl-D-aspartic acid ≥98% (by TLC)
Catalog # ENZOALX105009M050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)
  • Identity:
    Determined by NMR

Specification Test Results

About this item

 :
 :147.13 g/mol
Merck Index:14,06662
Storage Temperature:Ambient
CAS Number:6384-92-5
Formula:C₅H₉NO₄
MDL Number:MFCD00004226