Order Entry
Belgium
Orders LinkContactUsLinkComponent
Carbonate Borax Free Buffer Solution

Carbonate Borax Free Buffer Solution

Supplier:   REAGECON
Carbonate Borax Free Buffer Solution
Supplier:  REAGECON
Certificates

2 Options Available Below

Product Details & Documents

This range of Reagecon pH Buffers are sodium tetraborate (Borax) free. They are not subject to classification by EU regulatory authorities as toxic to reproduction under CLP category 1B. Similar in approach to the existing range of Reagecon Mercury Free Buffers, these Buffer Solutions have been developed to enhance user choice and allay safety concerns, without any impact on fitness for purpose or product performance.

  • Non toxic and borax free
  • Reagecon pH buffers are pre-programmed into the instruments of most major manufacturers
  • Laboratory chemical

For increased confidence in their test measurements, analysts should regularly measure the pH of a control standard. If an acceptable value is obtained from the control standard measurement then the analysts can have improved confidence that their test measurements will be correct. Reagecon’s extensive range of pH Buffers means that there will be a Reagecon pH Buffer which can be used as a control buffer for all pH applications.

Manufactured to exacting specifications with an extended shelf life and stability, even after opening the bottle thereby eliminating the requirement to open a fresh bottle of solution every time it is used.

Certifications: Reagecon’s Borax free pH buffer solutions are tested by our ISO 17025 accredited test method and NIST traceable.

Packaging: HDPE bottles.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • REGE1090BF
  • REGE110025BF
  • Physical form:
  • Liquid
  • Liquid

Specifications

Product Family Options

Product Information

  • pH value
    pH accuracy
    pH buffer type
    Reference temperature
    Pack type
    Pk
    Availability
    Price
  • 9,00
    ±0,01
    Carbonate
    20 °C
    HDPE bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 62,70 €
    62,70 €
    1 * 1 L
       62,70 €

    Specifications:
    More
    Specifications
    Cat. No.
    REGE1090BF
    Physical form
    Liquid
  • 10,00
    ±0,01
    Carbonate
    25 °C
    HDPE bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 66,10 €
    66,10 €
    1 * 1 L
       66,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    REGE110025BF
    Physical form
    Liquid
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.