Order Entry
Belgium
ContactUsLinkComponent
Z-Phe-Tyr(tBu)-Diazomethylketone
Z-Phe-Tyr(tBu)-Diazomethylketone
Supplier:  ENZO LIFE SCIENCES
Certificates
Z-Phe-Tyr(tBu)-Diazomethylketone
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Storage conditions:
    −20 °C
  • Protein/peptide name:
    Z-Phe-Tyr(tBu)-diazomethylketone
  • Molecular weight:
    542.63
  • Molecular formula:
    C₃₁H₃₄N₄O₅

Specifications

Frequently Bought Together