Orders LinkContactUsLinkComponent
Height Adjustable Workstations
Height Adjustable Workstations
Supplier:   Palbam
Height Adjustable Workstations
Supplier:  Palbam
Certificates
Height Adjustable Workstations
Height Adjustable Workstations
3 Options Available Below
Product Details & Documents
The cleanroom height adjustable workstation is designed with motorised lifters to allow the operator to quickly change the height of the table to ensure a comfortable, ergonomic work environment.
Table top and leveler base are manufactured from stainless steel 304 with electro-polished finish. The adjustable legs are manufactured from anodised aluminum. Height adjustability is controlled from easy to use front mounted control unit.
Table height can be adjusted from 760 to 1147 mm (30 to 45"). Custom dimensions are possible.
These tables are designed for 220 V.
Product Family Options
Product Information
  • Material
    Availability
    Price
  • PALBWU1001
    Cleanroom height adjustable workstation, 1000×600×767-1147 mm
    Cleanroom height adjustable workstation, 1000×600×767-1147 mm
    Stainless steel 304
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 720,00 €
    7 720,00 €
    1 * 1 ST
       7 720,00 €
  • PALBWU1201
    Cleanroom height adjustable workstation, 1200×600×767-1147 mm
    Cleanroom height adjustable workstation, 1200×600×767-1147 mm
    Stainless steel 304
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 220,00 €
    8 220,00 €
    1 * 1 ST
       8 220,00 €
  • PALBWU1401
    Cleanroom height adjustable workstation, 1400×600×767-1147 mm
    Cleanroom height adjustable workstation, 1400×600×767-1147 mm
    Stainless steel 304
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 700,00 €
    8 700,00 €
    1 * 1 ST
       8 700,00 €