BalanCD™ CHO Feed 3, Powder
Catalog # SHBT94118-10L
Supplier: Shenandoah Biotechnology
CAS Number:
Specifications
- Format du milieu:Powder
- Cdt:10 L
- Seq:01
- Taille:10 L
- Température de stockage:2...8 °C
- Durée de vie:24 mois
Specifications
About this item
BalanCD™ CHO Feed 3 is a chemically-defined, animal component-free feed medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells in fed-batch mode.
This formulation was developed using FUJIFILM Irvine Scientific’s Rational Culture Media Design strategy to achieve enhanced performance of growth and production in fed-batch culture. BalanCD™ CHO Feed 3 was designed to be used with a wide range of growth media for optimal performance in fed-batch cultures.