Screw Cap GL 40
Fournisseur: RIXIUS
New Product
Certificats
À propos de cet article
Screw cap made of black PP with ALU laminated PE foam insert for thread GL 40.
- Height: 14,4 mm
- Weight: 6 g
The PP screw caps makes it very popular for packaging food and cosmetics, among other things.
Frequently Bought Together