Order Entry
Belgium
ContactUsLinkComponent
Perfringolysin O, Recombinant, Clostridium Perfringens, aa29-500, His-Tag (PFO)
Perfringolysin O, Recombinant, Clostridium Perfringens, aa29-500, His-Tag (PFO)
Supplier:  US Biological
Certificates
Perfringolysin O, Recombinant, Clostridium Perfringens, aa29-500, His-Tag (PFO)
Supplier:  US Biological

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    C. perfringens
  • Storage conditions:
    −20 °C
  • Form:
    Supplied as a liquid in tris, 50% glycerol
  • Shipping conditions:
    20
  • Purity:
    ca.85% (SDS-PAGE)
  • Molecular weight:
    67, 92 (non Ab)
  • Grade:
    Purified

Specifications

Frequently Bought Together