Orders LinkContactUsLinkComponent
Mob Cap Detect, PP
Mob Cap Detect, PP
Blue, 500 mm, Cardboard (Outer); PE (Inner)
Catalog # MENS1216
Supplier:   MENSCH FRANZ
Mob Cap Detect, PP
Catalog # MENS1216
Supplier:  MENSCH FRANZ
Mob Cap Detect, PP
Blue, 500 mm, Cardboard (Outer); PE (Inner)
Price
82,80 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Detectable mob caps Detect made of PP by Hygostar are machine-made and therefore very inexpensive.
  • Breathable spunbonded PP
  • Detectable, machine-made
  • Latex-free elastic band
  • Comfortable to wear
  • Suitable for contact with food
  • Protects against low superficial mechanical risks
They are made of breathable PP fleece and are comfortable to wear in everyday work. The caps are detectable because they have an integrated metal strip. This metal strip can be detected by the detector machine. Detectors are mainly used in large food and pharmaceutical production facilities.
Like sniffer noses, they detect when a foreign object, such as a piece of clothing, is present in the production process. In this way, these machines ensure that, on the one hand, the end consumer receives a hygienically perfect product. On the other hand, the production plant is safe from damage caused by foreign bodies.
Functional range are food industry and hygiene sectors.
Certifications: PSA VO (EU) 2016-425, CAT 1 -  Personal protective equipment category 1, food safe and detectable.
Specifications
  • Ø:
    500 mm
  • Packed:
    Cardboard (Outer); PE (Inner)
  • Colour:
    Blue