Orders LinkContactUsLinkComponent
Storage Cleaners Cabinets
 :   Palbam
Storage Cleaners Cabinets
 :  Palbam
Storage Cleaners Cabinets
Designed for storing mops and cleaning materials.
  • Two doors
Fabrication material stainless steel 304 with electropolished finish.
Product Family Options
  • Description
    Doors
    Material
    W×D×H
  • Cleaners cabinet on casters
    2
    Stainless steel 304
    500×800×1800 mm
    1 * 1 ST
    1 * 1 ST
  • Cleaners cabinet on leveler legs
    2
    Stainless steel 304
    500×800×1800 mm
    1 * 1 ST
    1 * 1 ST