Orders LinkContactUsLinkComponent
Optical DO Sensor Cable
Optical DO Sensor Cable
Optical DO sensor cable
Catalog # DISI2700-0418
Supplier:   DISTEK
Optical DO Sensor Cable
Catalog # DISI2700-0418
Supplier:  DISTEK
Optical DO Sensor Cable
Optical DO sensor cable
Price
653,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Optical DO sensor cable that will work only with the optical DO sensors such as the Hamilton VisiFerm ECS optical DO sensor.
Specifications
  • Description:
    Optical DO sensor cable