Order Entry
Belgium
ContactUsLinkComponent
WHEATON® I-Loc™ Polypropylene, Screw Thread, Closure, with, Stopper
WHEATON® I-Loc™ Polypropylene, Screw Thread, Closure, with, Stopper
Catalog # WHEAW010342J
Supplier:  DWK Life Sciences
WHEATON® I-Loc™ Polypropylene, Screw Thread, Closure, with, Stopper
Catalog # WHEAW010342J
Supplier:  DWK Life Sciences

Specifications

  • Thread:
    20-400
  • Colour:
    Black
  • Material:
    PP
  • Pk:
    2.500

Specifications

About this item

For use with screw thread bottles and vials.

  • Siliconized grey bromobutyl stopper, 50 Durometer
  • Autoclavable

Polypropylene open top screw closure for piercing applications.