Foetal bovine serum, characterised, US origin, HyClone™
Supplier: HyClone products (Cytiva)
Certificates
About this item
Enhance growth of cells with U.S.-sourced Cytiva HyClone™ Characterized Fetal Bovine Serum (FBS). More economical than Defined FBS, but meets the requirements of most discriminating cell culturists.
Tested for endotoxin, hemoglobin, and additional biochemical assays and filtered through serial 100 nm pore-size rated filters.
Specifications
- Catalog No:
- SH30071.02
- SH30071.03IR25
- SH30071.03HI
- HYCLSH30071.01
- SH30071.03IR30
- SH30071.01HI
- HYCLSH30071.02HI
- SH30071.03
- Concentration:
- Endotoxin: ≤25 EU/ml
Hemoglobin: ≤25 mg/dl - Endotoxin: ≤25 EU/ml
Hemoglobin: ≤25 mg/dl - Endotoxin: ≤25 EU/ml
Hemoglobin: ≤25 mg/dl - Endotoxin: ≤25 EU/ml
Hemoglobin: ≤25 mg/dl - Endotoxin: ≤25 EU/ml
Hemoglobin: ≤25 mg/dl - Endotoxin: ≤25 EU/ml
Hemoglobin: ≤25 mg/dl - Endotoxin: ≤25 EU/ml
Hemoglobin: ≤25 mg/dl - Endotoxin: ≤25 EU/ml
Hemoglobin: ≤25 mg/dl
- Storage temperature:
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
Specifications
Related Information
Certificates
Search by Lot NumberCytiva HyClone™ Sera Fetal Bovine Serum, Characterised, US Origin
Product Literature
Frequently Bought Together