Drosophila containers
Supplier: Greiner Bio-One
Jump to:
About this item
PS, crystal-clear.
- For single use only
- Temperature resistance –20 to +60 °C
- Without Ceaprene stopper (must be ordered separately)
Ideally suited for the cultivation of Drosophila melanogaster.
2 Options Available Below
Product Details & Documents
PS, crystal-clear.
- For single use only
- Temperature resistance –20 to +60 °C
- Without Ceaprene stopper (must be ordered separately)
Ideally suited for the cultivation of Drosophila melanogaster.
Product Family Options
Product Information
Stopper
Product Information
- Catalog #Ø inférieurHauteurØ supérieurCouleurTypeAvailabilityPrice