Orders LinkContactUsLinkComponent
xPIPETTE™ Carousel Stand
xPIPETTE™ Carousel Stand
Carousel stand, For xPIPETTE™ pipettes, PP
Catalog # 613-6912
Supplier:   BIOTIX
xPIPETTE™ Carousel Stand
Catalog # 613-6912
Supplier:  BIOTIX
xPIPETTE™ Carousel Stand
Carousel stand, For xPIPETTE™ pipettes, PP
Price
411,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The carousel pipette stand is a rotating carousel stand for the xPIPETTE.
  • Fits both single and multichannel models
  • Holds seven pipettes total
  • Made from chemically resistant polypropylene
Pipettes hang vertically, preventing liquids from contaminating the pipette shaft.
Specifications
  • For:
    For xPIPETTE™ pipettes
  • Description:
    Carousel stand
  • Material:
    PP