Lead(II) acetate
HDPE Bottle, 500 mL
Catalog # REGEUSP3105
Supplier: REAGECON
Jump to:
Product Details & Documents
Certifications: ISO 17025 Accredited Test Method (A2LA Ref: 6739.03).
Synonyms: Acetic acid Lead(II) salt
- CAS Number:301-04-2
- Formula:(H₃CCOO)₂Pb
- MDL Number:MFCD00012452
- Molecular Weight:325.29 g/mol
- UN:3082
- ADR:9,III
Specifications
Specifications
- Pack type:HDPE Bottle
- Pk:500 mL



