Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 1.2 mol/l (1.2 N) in aqueous solution
Corrosive
Sodium hydroxide 1.2 mol/l (1.2 N) in aqueous solution
Catalog # KRAF06188.7900
Supplier:  Bernd Kraft
CAS Number:  
Corrosive
Sodium hydroxide 1.2 mol/l (1.2 N) in aqueous solution
Catalog # KRAF06188.7900
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 L
  • Seq:
    5327

Specifications

About this item

 :
 :40 g/mol
Density:1.04 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:3266
ADR:8,III
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548