
Sodium hydroxide 0.33 mol/l (1/3 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Catalog # 1.05595.9010
Supplier: Merck
CAS Number:
Specifications
- Pack type:PE/alu Titripac®
- Pk:10 L
- Seq:5214
Specifications
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.3332 - 0.3350
- Measurement uncertainty:+/- 0.0010
- Traceability:NIST SRM
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.02 g/cm³ (25 °C)
Storage Temperature:Ambient
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:215-185-5
UN:1824
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548