Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.05 M NaOH (0.05N) volumetric solution, Fluka™
Corrosive
Sodium hydroxide 0.05 M NaOH (0.05N) volumetric solution, Fluka™
Certificates
Corrosive
Sodium hydroxide 0.05 M NaOH (0.05N) volumetric solution, Fluka™
Certificates

Specification Test Results

  • Vapor Pressure:
    3 mmHg ( 37 °C)
  • Factor:
    0.995 - 1.005
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

Related Information

Frequently Bought Together