Order Entry
Belgium
Orders LinkContactUsLinkComponent
Copper, powder

Copper, powder

Copper, powder
Certificates
Copper, powder

3 Options Available Below

Product Details & Documents
  • CAS Number:7440-50-8
  • Formula:Cu
  • MDL Number:MFCD00010965
  • Molecular Weight:63.55 g/mol
  • Boiling Pt:2567 °C (1013 hPa)
  • Melting Pt:1083.4 °C
  • Density:8.94 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-159-6
  • UN:3089
  • ADR:4.1,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.