Order Entry
Belgium
ContactUsLinkComponent
Heptafluorobutyric anhydride (HFAA)
Heptafluorobutyric anhydride (HFAA)
Catalog # ROTH0026.1
Supplier:  Roth Carl
CAS Number:  
Heptafluorobutyric anhydride (HFAA)
Catalog # ROTH0026.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :410.06 g/mol
Boiling Pt:108 °C (1013 hPa)
Melting Pt:14 °C
Density:1.653 g/cm³ (25 °C)
EINECS:206-410-8
CAS Number:336-59-4
Formula:C₈F₁₄O₃
MDL Number:MFCD00000432