Non-Contact Dispensers, I.DOT
Fournisseur: DISPENDIX
Certificats
À propos de cet article
An accessible research appliance enabling the dispensing of nanoliter and microliter scale volumes to 96-, 384-, and 1536-well plates.
- Use a fraction of the pipette tips required by traditional automation
- Dispense reagents with a 1 μl dead volume and run more samples for the same reagent cost
- Dispense 4 nl to 30 μl into any SBS format destination labware with a high degree of precision
- Dispense with insight into the actual volume dispensed every time with integrated volume verification
- Software for dispensing a constant volume, dilution, normalization, and pooling should not be complicated
- Dispense up to 384 source liquids per run at a speed and precision that will continue to surprise with every use
The 'I.DOT' is a liquid handler that anyone can use regardless of automation experience. The instrument transfers volumes as low as 4 nl with unrivalled precision and speed, while dramatically reducing the laboratory’s plastics and reagent consumption.
Whether dispensing enzymes and beads for NGS or qPCR or adding cells, compounds, and buffers for Assay development or high-throughput screening, the 'I.DOT' offers unrivalled precision and ease of use for efficient and reproducible sample preparation.
Frequently Bought Together