Order Entry
Belgium
Orders LinkContactUsLinkComponent
Casein

Casein

Plastic bottle for solids, 100 g
Catalog # 1.02244.0100
Supplier:   Merck
Casein
Catalog # 1.02244.0100
Supplier:  Merck
CAS Number:  
Casein

Plastic bottle for solids, 100 g

CAS Number:  
Price
149,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Cat. No.:
    1.02244.0100
  • Protein/peptide name:
    Casein
  • Storage conditions:
    Ambient
  • CAS No.:
    9000-71-9

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.