BenchBin™ Benchtop Biohazard Bin
Catalog # ISMC14003-04
Supplier: ISMATEC PART OF COLE PARMER
Specifications
- Capacity:1,5 L
- Colour:Red
- Material:PP
- W×D×H:180×130×180 mm
- Pk:1
Specifications
About this item
The BenchBin™ is a compact, convenient, and safe biohazard bin for the benchtop or workstation.
- Push to open/close lid keeps waste materials covered
- Use with 216×279 mm disposal bags
- Compatible with autoclavable biohazard bags
Benchtop biohazard bin only measures 180L×130W×180H mm taking up minimal countertop space. Removable cover makes it quick and easy to empty contents.