ContactUsLinkComponent
Galtek® Check Valves, ³/₄" Orifice, O-ring Free Design E
Galtek® Check Valves, ³/₄" Orifice, O-ring Free Design E
Supplier:  ENTEGRIS
New Product
Certificates
Galtek® Check Valves, ³/₄" Orifice, O-ring Free Design E
Supplier:  ENTEGRIS
New Product
Specifications
  • Catalog No:
  • ENTECKM12-12F-12F
  • ENTECKM12-12K-12K
  • Length:
  • 113 mm
  • 113 mm
  • Max. pressure:
  • 690 kPa up to 65 °C
  • 690 kPa up to 65 °C
  • Max. temperature:
  • –18...+100 °C
  • –18...+100 °C
  • Wetted materials:
  • Molded PFA and PTFE
  • Molded PFA and PTFE