Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cholesterol

Cholesterol

Glass bottle for solids, 100 g
Catalog # A0807.0100
Supplier:   PanReac AppliChem
Cholesterol
Catalog # A0807.0100
Supplier:  PanReac AppliChem
CAS Number:  57-88-5
Cholesterol

Glass bottle for solids, 100 g

CAS Number:  57-88-5
Price
102,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
  • CAS Number:57-88-5
  • Formula:C₂₇H₄₆O
  • MDL Number:MFCD00003646
  • Molecular Weight:386.66 g/mol
  • Merck Index:13,02221
  • Boiling Pt:360 °C (1013 hPa)
  • Melting Pt:147…150 °C
  • Density:1.07 g/cm³ (20 °C)
  • Storage Temperature:Freezer
  • EINECS:200-353-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.